Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-Gly-Gly-Phe-Gly-PAB-OH (Mc-GGFG-PAB-OH) | 2632342-05-1 | 98.8% | 634.68 g/mol | C32H38N6O8 | 5 MG
SDP

Catalog No. 5000203871
Encompass_Preferred

Mc-Gly-Gly-Phe-Gly-PAB-OH (Mc-GGFG-PAB-OH) is a cleavable peptide-based antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies in ADC design. It is supplied as a solid, high-purity material suitable for research and linker development.

  • Cleavable peptide-based linker for ADC synthesis
  • High purity suitable for research applications
  • Solid form, white to yellow powder
  • Molecular weight approximately 634.68 g/mol
  • Storage: powder -80°C for long-term stability

Catalog No. 50-002-03871 Supplier Medchemexpress LLC Supplier No. HY1364325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mc-Gly-Gly-Phe-Gly-PAB-OH (Mc-GGFG-PAB-OH) is a cleavable peptide-based antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies in ADC design. It is supplied as a solid, high-purity material suitable for research and linker development.

  • Cleavable peptide-based linker for ADC synthesis
  • High purity suitable for research applications
  • Solid form, white to yellow powder
  • Molecular weight approximately 634.68 g/mol
  • Storage: powder -80°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.