Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-gly-gly-phe-gly-pab-oh TFA | 2632342-05-1 | 97.5% | 748.70 g/mol | C34H39F3N6O10 | 10 MG
SDP

Catalog No. 5000202710
Encompass_Preferred

Mc-Gly-Gly-Phe-Gly-PAB-OH TFA is the trifluoroacetate salt of a cleavable peptide linker used to assemble antibody-drug conjugates (ADCs). It provides a protease- or chemically cleavable spacer between an antibody and a cytotoxic payload, and is supplied for research use in small-scale packages.

  • Cleavable peptide linker suitable for ADC construction.
  • Provided as a TFA salt to improve solubility and stability.
  • Available in multiple small-scale package sizes for research use.
  • High purity suitable for research applications.
  • Molecular weight 748.70 g/mol and formula C34H39F3N6O10.

Catalog No. 50-002-02710 Supplier Medchemexpress LLC Supplier No. HY136432A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mc-Gly-Gly-Phe-Gly-PAB-OH TFA is the trifluoroacetate salt of a cleavable peptide linker used to assemble antibody-drug conjugates (ADCs). It provides a protease- or chemically cleavable spacer between an antibody and a cytotoxic payload, and is supplied for research use in small-scale packages.

  • Cleavable peptide linker suitable for ADC construction.
  • Provided as a TFA salt to improve solubility and stability.
  • Available in multiple small-scale package sizes for research use.
  • High purity suitable for research applications.
  • Molecular weight 748.70 g/mol and formula C34H39F3N6O10.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.