Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MC-Gly-Gly-Phe-Gly-PAB-PNP | 2632342-16-4 | 99.2% | 799.78 | C39H41N7O12 | 10 MG
SDP

Catalog No. 5000203402
Encompass_Preferred

MC-Gly-Gly-Phe-Gly-PAB-PNP is a protease-cleavable tetrapeptide linker supplied for research use and intended for use in the synthesis of antibody-drug conjugates targeting human EGFR.

  • Protease-cleavable tetrapeptide linker suitable for ADC synthesis.
  • Molecular formula C39H41N7O12.
  • Molecular weight 799.78 g/mol.
  • Purity 99.22%.
  • Soluble in DMSO at 100 mg/mL (125.03 mM) with sonication.
  • Supplied as a 10 MG vial for research applications.

Catalog No. 50-002-03402 Supplier Medchemexpress LLC Supplier No. HY14716410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-Gly-Gly-Phe-Gly-PAB-PNP is a protease-cleavable tetrapeptide linker supplied for research use and intended for use in the synthesis of antibody-drug conjugates targeting human EGFR.

  • Protease-cleavable tetrapeptide linker suitable for ADC synthesis.
  • Molecular formula C39H41N7O12.
  • Molecular weight 799.78 g/mol.
  • Purity 99.22%.
  • Soluble in DMSO at 100 mg/mL (125.03 mM) with sonication.
  • Supplied as a 10 MG vial for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.