Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-Gly-Gly-Phe-Gly-(R)-cyclopropane-exatecan | 2414254-51-4 | 97.4% | 1074.12 | 5 MG
SDP

Catalog No. 5000362428
Encompass_Preferred

Mc-Gly-Gly-Phe-Gly-(R)-Cyclopropane-Exatecan (SHR-A1811 Drug-linker) is an agent-linker conjugate for Antibody-Drug Conjugates (ADC), consisting of Exatecan. Exatecan is a DNA Topoisomerase I inhibitor (IC50=2.2 μM).

  • Contains Exatecan, a DNA Topoisomerase I inhibitor.
  • Solid, white to light yellow appearance.
  • Soluble in DMSO (100 mg/mL).
  • In vivo solubility in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (≥ 2.5 mg/mL) or 10% DMSO, 90% Corn Oil (≥ 2.5 mg/mL).
  • Store at -20°C, sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • For research use only.

Catalog No. 50-003-62428 Supplier Medchemexpress LLC Supplier No. HY1486685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mc-Gly-Gly-Phe-Gly-(R)-Cyclopropane-Exatecan (SHR-A1811 Drug-linker) is an agent-linker conjugate for Antibody-Drug Conjugates (ADC), consisting of Exatecan. Exatecan is a DNA Topoisomerase I inhibitor (IC50=2.2 μM).

  • Contains Exatecan, a DNA Topoisomerase I inhibitor.
  • Solid, white to light yellow appearance.
  • Soluble in DMSO (100 mg/mL).
  • In vivo solubility in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (≥ 2.5 mg/mL) or 10% DMSO, 90% Corn Oil (≥ 2.5 mg/mL).
  • Store at -20°C, sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.