Learn More
Enzo Life Sciences Microcystin LR monoclonal antibody (MC10E7) (200µg)
SDP
Supplier:  Enzo Life Sciences ALX804320C200
Encompass_Preferred
Clone: MC10E7. Isotype: IgG1. Immunogen: Microcystin-LR linked via N-methyl-dehydroalanine to cationized ovalbumin. Species reactivity: Species independent. Specificity: Recognizes all 4-Arg microcystins. Applications: ELISA, IHC (PS), WB. Purity: Protein G-affinity purified. Formulation: Lyophilized.
Catalog No. 50-200-9861