Learn More
Medchemexpress LLC HY-15741 1mg , Mc-MMAE CAS:863971-24-8 Purity:>98%
SDP
Catalog No. 502260941
Encompass_Preferred
HY-15741 1mg Mc-MMAE CAS: 863971-24-8 Mc-MMAE is a protective group (maleimidocaproyl)-conjugated monomethyl auristatin E (MMAE), which is a potent tubulin inhibitor. Mc-MMAE is a drug-linker conjugate for ADC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0941 Supplier Medchemexpress LLC Supplier No. HY157411MG
HY-15741 1mg Mc-MMAE CAS: 863971-24-8 Mc-MMAE is a protective group (maleimidocaproyl)-conjugated monomethyl auristatin E (MMAE), which is a potent tubulin inhibitor. Mc-MMAE is a drug-linker conjugate for ADC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.