Learn More
Medchemexpress LLC Mc-peg2-va-pab-exatecan | 2845165-13-9 | 99.6% | 994.03 | C51H56FN7O13 | 10 MG
SDP
Catalog No. 5000287852
Encompass_Preferred
MC-PEG2-VA-PAB-Exatecan is an exatecan-based drug-linker conjugate designed for antibody-drug conjugate (ADC) research. It links the camptothecin-derived cytotoxic payload exatecan to a cleavable PEG2-Val-Cit-PAB linker to permit targeted intracellular release of the payload during ADC development.
  • Exatecan payload confers potent topoisomerase I inhibition.
  • Cleavable Val-Cit-PAB linker enables enzyme-triggered release inside target cells.
  • PEG2 spacer improves solubility and conjugation flexibility.
  • High reported purity supports reproducible research results.
  • Available in mg-scale quantities suitable for ADC synthesis workflows.
Catalog No. 50-002-87852 Supplier Medchemexpress LLC Supplier No. HY15674810MG
MC-PEG2-VA-PAB-Exatecan is an exatecan-based drug-linker conjugate designed for antibody-drug conjugate (ADC) research. It links the camptothecin-derived cytotoxic payload exatecan to a cleavable PEG2-Val-Cit-PAB linker to permit targeted intracellular release of the payload during ADC development.
  • Exatecan payload confers potent topoisomerase I inhibition.
  • Cleavable Val-Cit-PAB linker enables enzyme-triggered release inside target cells.
  • PEG2 spacer improves solubility and conjugation flexibility.
  • High reported purity supports reproducible research results.
  • Available in mg-scale quantities suitable for ADC synthesis workflows.