Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-PEG4-Val-Ala-PAB-Exatecan | 99.7% | 1153.21 | C58H69FN8O16 | 5 MG
SDP

Catalog No. 5000203310
Encompass_Preferred

Mc-PEG4-Val-Ala-PAB-Exatecan is an ADC drug-linker conjugate composed of a cleavable Val-Ala-PAB linker and the DNA topoisomerase I inhibitor Exatecan; supplied for research use in milligram quantities for ADC development and linker-payload studies.

  • Purity: 99.7%
  • Molecular weight: 1153.21
  • Chemical formula: C58H69FN8O16
  • Physical state: Solid
  • Available in 1 MG, 5 MG, 10 MG, 25 MG, 50 MG, 100 MG
  • Storage: -20°C sealed; in solvent -80°C (6 months), -20°C (1 month)
  • Solubility in DMSO: ≥ 100 mg/mL

Catalog No. 50-002-03310 Supplier Medchemexpress LLC Supplier No. HY1715805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mc-PEG4-Val-Ala-PAB-Exatecan is an ADC drug-linker conjugate composed of a cleavable Val-Ala-PAB linker and the DNA topoisomerase I inhibitor Exatecan; supplied for research use in milligram quantities for ADC development and linker-payload studies.

  • Purity: 99.7%
  • Molecular weight: 1153.21
  • Chemical formula: C58H69FN8O16
  • Physical state: Solid
  • Available in 1 MG, 5 MG, 10 MG, 25 MG, 50 MG, 100 MG
  • Storage: -20°C sealed; in solvent -80°C (6 months), -20°C (1 month)
  • Solubility in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.