Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-Pro-PAB-MMAE | 2766450-03-5 | 99.2% | 1214.49 | C64H95N9O14 | 10 MG
SDP

Catalog No. 5000203001
Encompass_Preferred

Mc-Pro-PAB-MMAE is a drug-linker conjugate used in the synthesis of antibody-drug conjugates (ADCs). It combines monomethyl auristatin E with a para-aminobenzyl (PAB)-type linker and is supplied as a solid for research use only.

  • Consists of monomethyl auristatin E and a PAB-type linker.
  • Molecular formula C64H95N9O14 and molecular weight 1214.49.
  • CAS number 2766450-03-5.
  • Purity approximately 99.2%.
  • White to off-white solid appearance.
  • Soluble in DMSO at ≥100 mg/mL.
  • Available in multiple milligram pack sizes for research applications.
  • Intended for research use only; not for human therapeutic use.

Catalog No. 50-002-03001 Supplier Medchemexpress LLC Supplier No. HY17106010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mc-Pro-PAB-MMAE is a drug-linker conjugate used in the synthesis of antibody-drug conjugates (ADCs). It combines monomethyl auristatin E with a para-aminobenzyl (PAB)-type linker and is supplied as a solid for research use only.

  • Consists of monomethyl auristatin E and a PAB-type linker.
  • Molecular formula C64H95N9O14 and molecular weight 1214.49.
  • CAS number 2766450-03-5.
  • Purity approximately 99.2%.
  • White to off-white solid appearance.
  • Soluble in DMSO at ≥100 mg/mL.
  • Available in multiple milligram pack sizes for research applications.
  • Intended for research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.