Learn More
Bellco Glass, Inc. MicroCarrier Screw Cap Assembly , 100mm Micro-carrier - 6-36L
SDP
Catalog No. 501945134
Encompass_Preferred
Micro-carrier screw cap assembly. •Fits center neck of 6L to 36L MicroCarrier Flasks. MADE IN THE USA•Includes: One 100mm blue PP screw cap with 0.77” hole, one 100mm white silicone liner with ¾” hole, one black PP compression fitting with 9mm hole
Catalog No. 50-194-5134 Supplier Bellco Glass, Inc. Supplier No. 196599100
Micro-carrier screw cap assembly. •Fits center neck of 6L to 36L MicroCarrier Flasks. MADE IN THE USA•Includes: One 100mm blue PP screw cap with 0.77” hole, one 100mm white silicone liner with ¾” hole, one black PP compression fitting with 9mm hole