Learn More
Bellco Glass, Inc. MicroCarrier Screw Cap Assembly , 70mm MicroCarrier 100-500mL
SDP
Catalog No. 501945105
Encompass_Preferred
Polypropylene screw cap with silicone gasket pouring ring and compression fitting. 70mm. MADE IN THE USA
Catalog No. 50-194-5105 Supplier Bellco Glass, Inc. Supplier No. 196590070
Polypropylene screw cap with silicone gasket pouring ring and compression fitting. 70mm. MADE IN THE USA