Learn More
Medchemexpress LLC Mc-va-pabc-mmae | 1818864-51-5 | 1230.53 | C65H99N9O14 | 1 MG
SDP
Catalog No. 5000202218
Encompass_Preferred
MC-VA-PABC-MMAE is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples a peptide-based linker to monomethyl auristatin E (MMAE), a potent tubulin polymerization inhibitor. It is intended for use in ADC assembly and preclinical characterization.
  • Peptide-based ADC linker coupled to a tubulin inhibitor payload.
  • Suitable for ADC assembly and preclinical characterization.
  • Supplied in milligram quantities for research applications.
  • Available in multiple sizes to support small-scale studies.
  • Provides defined linker-payload chemistry for conjugation workflows.
Catalog No. 50-002-02218 Supplier Medchemexpress LLC Supplier No. HY1472391MG
MC-VA-PABC-MMAE is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples a peptide-based linker to monomethyl auristatin E (MMAE), a potent tubulin polymerization inhibitor. It is intended for use in ADC assembly and preclinical characterization.
  • Peptide-based ADC linker coupled to a tubulin inhibitor payload.
  • Suitable for ADC assembly and preclinical characterization.
  • Supplied in milligram quantities for research applications.
  • Available in multiple sizes to support small-scale studies.
  • Provides defined linker-payload chemistry for conjugation workflows.