Learn More
Medchemexpress LLC Mc-val-ala-oh | 1342211-31-7 | MFCD27956998 | 99.8% | 381.42 g/mol | C18H27N3O6 | 25 MG
SDP
Catalog No. 5000212111
Encompass_Preferred
MC-Val-Ala-OH is a cleavable antibody-drug conjugate (ADC) linker that incorporates a valine-alanine dipeptide and a maleimide reactive handle. It acts as a protease-sensitive spacer to enable intracellular release of conjugated payloads and is provided as a high-purity solid for research use only.
  • Protease-sensitive val-ala dipeptide for intracellular cleavage
  • Maleimide reactive handle for thiol-selective conjugation
  • High purity suitable for research-grade ADC synthesis
  • Soluble in common organic solvents such as DMSO
  • Available in lab-scale quantities for small-batch synthesis
Catalog No. 50-002-12111 Supplier Medchemexpress LLC Supplier No. HY10115325MG
MC-Val-Ala-OH is a cleavable antibody-drug conjugate (ADC) linker that incorporates a valine-alanine dipeptide and a maleimide reactive handle. It acts as a protease-sensitive spacer to enable intracellular release of conjugated payloads and is provided as a high-purity solid for research use only.
  • Protease-sensitive val-ala dipeptide for intracellular cleavage
  • Maleimide reactive handle for thiol-selective conjugation
  • High purity suitable for research-grade ADC synthesis
  • Soluble in common organic solvents such as DMSO
  • Available in lab-scale quantities for small-batch synthesis