Learn More
Medchemexpress LLC Mc-val-ala-oh | 1342211-31-7 | MFCD27956998 | 99.8% | 381.42 g/mol | C18H27N3O6 | 250 MG
SDP
Catalog No. 5000211488
Encompass_Preferred
MC-Val-Ala-OH is a cleavable linker reagent used to assemble antibody-drug conjugates (ADCs). It contains a Val-Ala dipeptide spacer and a maleimide functionality for thiol-reactive conjugation, and is supplied for research use only.
  • Cleavable Val-Ala peptide linker enabling payload release under intracellular conditions.
  • Maleimide functional group for efficient thiol-specific conjugation to antibodies.
  • High reported purity (99.8%) supporting reproducible conjugation chemistry.
  • Available in multiple pack sizes, including a 250 mg option for scale flexibility.
  • Stable when stored sealed at -20°C; protect from moisture and light.
Catalog No. 50-002-11488 Supplier Medchemexpress LLC Supplier No. HY101153250MG
MC-Val-Ala-OH is a cleavable linker reagent used to assemble antibody-drug conjugates (ADCs). It contains a Val-Ala dipeptide spacer and a maleimide functionality for thiol-reactive conjugation, and is supplied for research use only.
  • Cleavable Val-Ala peptide linker enabling payload release under intracellular conditions.
  • Maleimide functional group for efficient thiol-specific conjugation to antibodies.
  • High reported purity (99.8%) supporting reproducible conjugation chemistry.
  • Available in multiple pack sizes, including a 250 mg option for scale flexibility.
  • Stable when stored sealed at -20°C; protect from moisture and light.