Learn More
Cayman Chemical Mc-Val-Cit-PAB, Purity: ≥98%, CAS: 159857-80-4, 50 mg
SDP
Catalog No. 502937149
Encompass_Preferred
A peptide linker molecule; used in the synthesis of ADCs; has been used in the synthesis of an ADC containing the tubulin polymerization inhibitor KGP05; has been used as a precursor in the synthesis of Mc-Val-Cit-PABC-PNP
Catalog No. 50-293-7149 Supplier Cayman Chemical Supplier No. 2828550 MG
A peptide linker molecule; used in the synthesis of ADCs; has been used in the synthesis of an ADC containing the tubulin polymerization inhibitor KGP05; has been used as a precursor in the synthesis of Mc-Val-Cit-PABC-PNP