Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MC-Val-Cit-PAB-Auristatin E | 2055896-77-8 | 97.0% | 1287.65 | C68H108N11O13 | 5 MG
SDP

Catalog No. 5000203387
Encompass_Preferred

MC-Val-Cit-PAB-Auristatin E is an antibody-drug conjugate (ADC) linker-payload conjugate for research use. It links the cytotoxic tubulin inhibitor auristatin E to an MC-Val-Cit-PAB cleavable linker, enabling enzymatic release of the payload in target cells. The material is provided as a solid for preclinical and ADC development studies.

  • cleavable val-cit dipeptide linker enabling enzymatic payload release
  • contains auristatin E tubulin inhibitor as the payload
  • molecular formula C68H108N11O13 and molecular weight 1287.65
  • purity ~97.0% by HPLC (batch-specific certificate of analysis available)
  • supplied as a solid with good solubility in DMSO for formulation

Catalog No. 50-002-03387 Supplier Medchemexpress LLC Supplier No. HY1288995MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-Val-Cit-PAB-Auristatin E is an antibody-drug conjugate (ADC) linker-payload conjugate for research use. It links the cytotoxic tubulin inhibitor auristatin E to an MC-Val-Cit-PAB cleavable linker, enabling enzymatic release of the payload in target cells. The material is provided as a solid for preclinical and ADC development studies.

  • cleavable val-cit dipeptide linker enabling enzymatic payload release
  • contains auristatin E tubulin inhibitor as the payload
  • molecular formula C68H108N11O13 and molecular weight 1287.65
  • purity ~97.0% by HPLC (batch-specific certificate of analysis available)
  • supplied as a solid with good solubility in DMSO for formulation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.