Learn More
Medchemexpress LLC MC-Val-Cit-PAB-Auristatin E | 2055896-77-8 | 97.0% | 1287.65 g/mol | C68H108N11O13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1288991MG
Encompass_Preferred
MC-Val-Cit-PAB-Auristatin E is an antibody-drug conjugate (ADC) drug-linker conjugate that links auristatin E to a cathepsin-cleavable MC-Val-Cit-PAB linker for use in ADC research and antitumor studies.
  • Contains auristatin E as the cytotoxic payload.
  • Features a cathepsin-cleavable Val-Cit-PAB linker for intracellular release.
  • High purity suitable for research applications.
  • Solid form with storage at 4°C to maintain stability.
  • Available in small milligram pack sizes for pilot studies.
Catalog No. 50-002-02092