Learn More
TARGETMOL CHEMICALS INC Mc-Val-Cit-PAB-Cl 1MG
SDP
Catalog No. 502876898
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mc-Val-Cit-PAB-Cl, a cleavable ADC linker, is utilized in the conjugation of MMAE and antibodies to produce compounds such as antibody-MC-vc-MMAE (e.g., anti-CD22-MC-VC-PABC-MMAE), which demonstrate cytotoxicity with IC50s of 3.3 and 0.95 nM against BJAB and WSU cell lines, respectively. Purity 99.79%
Catalog No. 50-287-6898 Supplier TARGETMOL CHEMICALS INC Supplier No. T183211MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mc-Val-Cit-PAB-Cl, a cleavable ADC linker, is utilized in the conjugation of MMAE and antibodies to produce compounds such as antibody-MC-vc-MMAE (e.g., anti-CD22-MC-VC-PABC-MMAE), which demonstrate cytotoxicity with IC50s of 3.3 and 0.95 nM against BJAB and WSU cell lines, respectively. Purity 99.79%