Learn More
Medchemexpress LLC MC-Val-Cit-PAB-dimethylDNA31 | 1639352-06-9 | 99.6% | 1518.15 | C78H101ClN10O19 | 1 MG
SDP
Catalog No. 5000204173
Encompass_Preferred
MC-Val-Cit-PAB-dimethylDNA31 is a linker-drug conjugate provided for research use in antibody-drug conjugate (ADC) development. It couples the dimethylDNA31 payload to a protease-cleavable Val-Cit-PAB linker with a maleimidocaproyl functionality, and is supplied as a solid for laboratory conjugation and characterization.
  • Provided for research use in ADC development and related studies.
  • Available in small milligram quantities suitable for early-stage conjugation experiments.
  • High reported purity for chemical characterization and analytical work.
  • Contains a protease-cleavable Val-Cit dipeptide linkage for intracellular payload release.
  • Supplied as a stable solid intended for conjugation to antibodies or other targeting moieties.
Catalog No. 50-002-04173 Supplier Medchemexpress LLC Supplier No. HY1289051MG
MC-Val-Cit-PAB-dimethylDNA31 is a linker-drug conjugate provided for research use in antibody-drug conjugate (ADC) development. It couples the dimethylDNA31 payload to a protease-cleavable Val-Cit-PAB linker with a maleimidocaproyl functionality, and is supplied as a solid for laboratory conjugation and characterization.
  • Provided for research use in ADC development and related studies.
  • Available in small milligram quantities suitable for early-stage conjugation experiments.
  • High reported purity for chemical characterization and analytical work.
  • Contains a protease-cleavable Val-Cit dipeptide linkage for intracellular payload release.
  • Supplied as a stable solid intended for conjugation to antibodies or other targeting moieties.