Learn More
Medchemexpress LLC HY-128904 5mg , MC-Val-Cit-PAB-duocarmycin (chloride) CAS:2055896-98-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1289045MG
Encompass_Preferred
HY-128904 5mg MC-Val-Cit-PAB-duocarmycin (chloride) CAS: 2055896-98-3 MC-Val-Cit-PAB-duocarmycin chloride is a drug-linker conjugate for ADC with potent antitumor activity by using Duocarmycin (a DNA minor groove binding alkylating agent), linked via the ADC linker MC-Val-Cit-PAB. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9236