Learn More
Medchemexpress LLC MC-Val-Cit-PAB-duocarmycin chloride | 2055896-98-3 | 99.0% | 1055.05 | C54H65Cl2N9O9 | 10 MG
SDP
Catalog No. 5000203620
Encompass_Preferred
MC-Val-Cit-PAB-duocarmycin chloride is an antibody-drug conjugate (ADC) agent-linker conjugate that links a valine-citrulline cleavable linker and para-aminobenzyl (PAB) spacer to a duocarmycin payload for use in ADC research. It is supplied as a solid, high-purity compound intended for evaluating linker-payload performance and cytotoxic activity in preclinical studies.
  • Cleavable valine-citrulline linker with para-aminobenzyl spacer.
  • Duocarmycin payload that binds the DNA minor groove and alkylates DNA.
  • High purity (99.0%) solid suitable for research applications.
  • Supplied as a 10 MG quantity.
  • Store at -80°C, protect from light; unstable in solution.
Catalog No. 50-002-03620 Supplier Medchemexpress LLC Supplier No. HY12890410MG
MC-Val-Cit-PAB-duocarmycin chloride is an antibody-drug conjugate (ADC) agent-linker conjugate that links a valine-citrulline cleavable linker and para-aminobenzyl (PAB) spacer to a duocarmycin payload for use in ADC research. It is supplied as a solid, high-purity compound intended for evaluating linker-payload performance and cytotoxic activity in preclinical studies.
  • Cleavable valine-citrulline linker with para-aminobenzyl spacer.
  • Duocarmycin payload that binds the DNA minor groove and alkylates DNA.
  • High purity (99.0%) solid suitable for research applications.
  • Supplied as a 10 MG quantity.
  • Store at -80°C, protect from light; unstable in solution.