Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-val-cit-pab-indibulin | 2055896-95-0 | 95.8% | 945.48 | C50H55ClN9O8 | 5 MG
SDP

Catalog No. 5000202383
Encompass_Preferred

MC-Val-Cit-PAB-Indibulin is an antibody-drug conjugate (ADC) agent-linker conjugate that couples the MC-Val-Cit-PAB cleavable linker to indibulin for use in ADC research. It is supplied as a solid for research use only and is intended as a building block or payload-linker component for in vitro and in vivo antitumor studies.

  • Agent-linker conjugate using indibulin as the cytotoxic payload.
  • Molecular formula: C50H55ClN9O8; molecular weight: 945.48.
  • High purity around 95.8% as provided by the supplier.
  • Physical form: solid, light yellow to brown.
  • Soluble in DMSO (125 mg/mL); example in vivo vehicle: 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline.
  • Storage: keep sealed away from moisture; -20°C recommended; in solvent, store at -80°C for long-term.
  • Intended for research use only and not for clinical or therapeutic use.

Catalog No. 50-002-02383 Supplier Medchemexpress LLC Supplier No. HY1289085MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-Val-Cit-PAB-Indibulin is an antibody-drug conjugate (ADC) agent-linker conjugate that couples the MC-Val-Cit-PAB cleavable linker to indibulin for use in ADC research. It is supplied as a solid for research use only and is intended as a building block or payload-linker component for in vitro and in vivo antitumor studies.

  • Agent-linker conjugate using indibulin as the cytotoxic payload.
  • Molecular formula: C50H55ClN9O8; molecular weight: 945.48.
  • High purity around 95.8% as provided by the supplier.
  • Physical form: solid, light yellow to brown.
  • Soluble in DMSO (125 mg/mL); example in vivo vehicle: 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline.
  • Storage: keep sealed away from moisture; -20°C recommended; in solvent, store at -80°C for long-term.
  • Intended for research use only and not for clinical or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.