Learn More
Medchemexpress LLC Mc-val-cit-pab-indibulin | 2055896-95-0 | 95.8% | 945.48 g/mol | C50H55ClN9O8 | 10 MG
SDP
Catalog No. 5000204118
Encompass_Preferred
MC-Val-Cit-PAB-Indibulin is an antibody-drug conjugate (ADC) drug-linker conjugate that links indibulin to a cleavable val-cit-pab linker for use in ADC research. It is provided as a purified research reagent for preclinical studies and ADC payload development.
  • contains an indibulin payload linked via a cleavable val-cit-pab spacer
  • intended for research use in ADC development and antitumor studies
  • high purity suitable for analytical and preclinical applications
  • available in small research quantities for conjugation and assay work
Catalog No. 50-002-04118 Supplier Medchemexpress LLC Supplier No. HY12890810MG
MC-Val-Cit-PAB-Indibulin is an antibody-drug conjugate (ADC) drug-linker conjugate that links indibulin to a cleavable val-cit-pab linker for use in ADC research. It is provided as a purified research reagent for preclinical studies and ADC payload development.
  • contains an indibulin payload linked via a cleavable val-cit-pab spacer
  • intended for research use in ADC development and antitumor studies
  • high purity suitable for analytical and preclinical applications
  • available in small research quantities for conjugation and assay work