Learn More
Medchemexpress LLC Mc-val-cit-pab-indibulin | 2055896-95-0 | MFCD32201121 | 95.8% | 945.49 g/mol | C50H55ClN9O8+ | 50 MG
SDP
Catalog No. 5000202639
Encompass_Preferred
MC-Val-Cit-PAB-Indibulin is an antibody-drug conjugate (ADC) drug-linker conjugate in which the tubulin inhibitor indibulin is attached via a cleavable MC-Val-Cit-PAB linker. It is intended for research use in ADC development and preclinical studies.
  • Provides an indibulin payload linked through a protease-cleavable Val-Cit spacer
  • Offers a maleimidocaproyl (MC) reactive handle for thiol conjugation
  • Suitable for antibody-drug conjugate synthesis and preclinical evaluation
  • Reported purity suitable for research applications
  • Available in multiple milligram-scale package sizes for development needs
Catalog No. 50-002-02639 Supplier Medchemexpress LLC Supplier No. HY12890850MG
MC-Val-Cit-PAB-Indibulin is an antibody-drug conjugate (ADC) drug-linker conjugate in which the tubulin inhibitor indibulin is attached via a cleavable MC-Val-Cit-PAB linker. It is intended for research use in ADC development and preclinical studies.
  • Provides an indibulin payload linked through a protease-cleavable Val-Cit spacer
  • Offers a maleimidocaproyl (MC) reactive handle for thiol conjugation
  • Suitable for antibody-drug conjugate synthesis and preclinical evaluation
  • Reported purity suitable for research applications
  • Available in multiple milligram-scale package sizes for development needs