Learn More
Medchemexpress LLC MC-Val-Cit-PAB-vinblastine | 2055896-92-7 | 99.5% | 1366.62 | C74H97N10O15 | 10 MG
SDP
Catalog No. 5000202072
Encompass_Preferred
MC-Val-Cit-PAB-vinblastine is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links the microtubule inhibitor vinblastine to a cleavable val-cit-PAB-type linker. It is supplied as a white to light brown solid (CAS 2055896-92-7), with molecular formula C74H97N10O15 and molecular weight 1366.62 g/mol. Intended for research and analytical applications; not for human therapeutic use.
  • Drug-linker conjugate designed for ADC research
  • Vinblastine payload with microtubule inhibitory activity
  • Cleavable linker for lysosomal release
  • High purity suitable for analytical applications
  • Solid form, white to light brown appearance
  • Storage recommendations: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month)
  • Available as a 10 mg pack
Catalog No. 50-002-02072 Supplier Medchemexpress LLC Supplier No. HY12890210MG
MC-Val-Cit-PAB-vinblastine is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links the microtubule inhibitor vinblastine to a cleavable val-cit-PAB-type linker. It is supplied as a white to light brown solid (CAS 2055896-92-7), with molecular formula C74H97N10O15 and molecular weight 1366.62 g/mol. Intended for research and analytical applications; not for human therapeutic use.
  • Drug-linker conjugate designed for ADC research
  • Vinblastine payload with microtubule inhibitory activity
  • Cleavable linker for lysosomal release
  • High purity suitable for analytical applications
  • Solid form, white to light brown appearance
  • Storage recommendations: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month)
  • Available as a 10 mg pack