Learn More
Medchemexpress LLC HY-20336 100mg , Mc-Val-Cit-PABC-PNP CAS:159857-81-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY20336100MG
Encompass_Preferred
HY-20336 100mg Mc-Val-Cit-PABC-PNP CAS: 159857-81-5 Mc-Val-Cit-PABC-PNP is a cathepsin cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1064