Learn More
Cayman Chemical Mc-Val-Cit-PABC-PNP, Purity: ≥98%, CAS: 159857-81-5, 1 mg
SDP
Catalog No. 502994465
Encompass_Preferred
A peptide linker molecule; used in the synthesis of antibody-drug conjugates
Catalog No. 50-299-4465 Supplier Cayman Chemical Supplier No. 238811 MG
A peptide linker molecule; used in the synthesis of antibody-drug conjugates