Learn More
Medchemexpress LLC MC-VC-PAB-NH2 TFA | 1616727-21-9 | 99.9% | 772.77 | 1 MG
SDP
Catalog No. 5000348733
Encompass_Preferred
MC-VC-PAB-NH2 TFA is a cleavable antibody-drug conjugate (ADC) linker, utilized in the synthesis of ADCs. ADCs are composed of an antibody to which an ADC cytotoxin is attached via an ADC linker.
  • Cleavable antibody-drug conjugate linker
  • Utilized in the synthesis of ADCs
  • High purity of 99.9%
  • Appears as a white to off-white solid
  • Protease cleavable
Catalog No. 50-003-48733 Supplier Medchemexpress LLC Supplier No. HY136132A1MG
MC-VC-PAB-NH2 TFA is a cleavable antibody-drug conjugate (ADC) linker, utilized in the synthesis of ADCs. ADCs are composed of an antibody to which an ADC cytotoxin is attached via an ADC linker.
  • Cleavable antibody-drug conjugate linker
  • Utilized in the synthesis of ADCs
  • High purity of 99.9%
  • Appears as a white to off-white solid
  • Protease cleavable