Learn More
Medchemexpress LLC MC-VC-PAB-NH2 TFA | 1616727-21-9 | MFCD00935992 | 99.9% | 772.77 | C33H47F3N8O10 | 5 MG
SDP
Catalog No. 5000221810
Encompass_Preferred
MC-VC-PAB-NH2 TFA is a cleavable antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies and enable intracellular release through enzymatic cleavage. It contains a valine-citrulline dipeptide and a para-aminobenzyl self-immolative spacer and is supplied as the trifluoroacetate salt for stability in research applications.
  • Cleavable valine-citrulline linker for cathepsin-mediated payload release.
  • Para-aminobenzyl self-immolative spacer to facilitate efficient drug release.
  • Supplied as a trifluoroacetate salt for improved stability and handling.
  • High purity suitable for research use.
  • Soluble in DMSO at high concentration; ultrasonic assistance may be required.
  • Store sealed, away from moisture and light; follow recommended low-temperature storage for solutions.
Catalog No. 50-002-21810 Supplier Medchemexpress LLC Supplier No. HY136132A5MG
MC-VC-PAB-NH2 TFA is a cleavable antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies and enable intracellular release through enzymatic cleavage. It contains a valine-citrulline dipeptide and a para-aminobenzyl self-immolative spacer and is supplied as the trifluoroacetate salt for stability in research applications.
  • Cleavable valine-citrulline linker for cathepsin-mediated payload release.
  • Para-aminobenzyl self-immolative spacer to facilitate efficient drug release.
  • Supplied as a trifluoroacetate salt for improved stability and handling.
  • High purity suitable for research use.
  • Soluble in DMSO at high concentration; ultrasonic assistance may be required.
  • Store sealed, away from moisture and light; follow recommended low-temperature storage for solutions.