Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MC-VC-PAB-NH2 trifluoroacetic acid | 1616727-21-9 | 99.9% | C33H47F3N8O10 | 10MG
SDP

Catalog No. 5000259903
Encompass_Preferred

MC-VC-PAB-NH2 TFA is a cleavable ADC linker supplied as the trifluoroacetic acid salt for research use. It contains a Val-Cit-PAB cleavable spacer designed for protease-triggered drug release and is provided as a white to off-white solid with high purity.

  • Provides a protease-cleavable Val-Cit-PAB spacer for controlled payload release.
  • Supplied as a TFA salt to aid handling and stability.
  • High purity (≈99.9%) suitable for conjugation workflows.
  • Soluble in DMSO at high concentration (100 mg/mL) with ultrasonic assistance.
  • Solid, white to off-white appearance for convenient storage and handling.
  • Storage recommendations: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-59903 Supplier Medchemexpress LLC Supplier No. HY136132A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-VC-PAB-NH2 TFA is a cleavable ADC linker supplied as the trifluoroacetic acid salt for research use. It contains a Val-Cit-PAB cleavable spacer designed for protease-triggered drug release and is provided as a white to off-white solid with high purity.

  • Provides a protease-cleavable Val-Cit-PAB spacer for controlled payload release.
  • Supplied as a TFA salt to aid handling and stability.
  • High purity (≈99.9%) suitable for conjugation workflows.
  • Soluble in DMSO at high concentration (100 mg/mL) with ultrasonic assistance.
  • Solid, white to off-white appearance for convenient storage and handling.
  • Storage recommendations: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.