Learn More
Medchemexpress LLC MC-VC-PABC-amide-PEG1-CH2-CC-885 | 2722697-82-5 | 99.9% | C55H68ClN11O13 | 1 MG
SDP
Catalog No. 5000354141
Encompass_Preferred
MC-VC-PABC-amide-PEG1-CH2-CC-885 is a neoDegrader-Linker conjugate consisting of the GSPT1 degrader/molecular glue CC-885 and a linker. This compound is utilized in the synthesis of Antibody-Drug Conjugates (ADCs) and Antibody neoDegrader Conjugates (AnDCs), which are designed to specifically degrade intracellular target proteins within target cells. It can be conjugated to various antibodies to form these conjugates.
  • Consists of a GSPT1 degrader/molecular glue and a linker.
  • Can be conjugated to various antibodies to form conjugates.
  • Designed to specifically degrade intracellular target proteins within target cells.
  • Utilized in the synthesis of antibody-drug conjugates (ADCs).
  • Utilized in the synthesis of antibody neoDegrader conjugates (AnDCs).
Catalog No. 50-003-54141 Supplier Medchemexpress LLC Supplier No. HY1454481MG
MC-VC-PABC-amide-PEG1-CH2-CC-885 is a neoDegrader-Linker conjugate consisting of the GSPT1 degrader/molecular glue CC-885 and a linker. This compound is utilized in the synthesis of Antibody-Drug Conjugates (ADCs) and Antibody neoDegrader Conjugates (AnDCs), which are designed to specifically degrade intracellular target proteins within target cells. It can be conjugated to various antibodies to form these conjugates.
  • Consists of a GSPT1 degrader/molecular glue and a linker.
  • Can be conjugated to various antibodies to form conjugates.
  • Designed to specifically degrade intracellular target proteins within target cells.
  • Utilized in the synthesis of antibody-drug conjugates (ADCs).
  • Utilized in the synthesis of antibody neoDegrader conjugates (AnDCs).