Learn More
Medchemexpress LLC MC-VC-PABC-C6-α-amanitin | 2922514-69-8 | 98.9% | 1616.79 g/mol | C74H105N17O22S | 1 MG
SDP
Catalog No. 5000202426
Encompass_Preferred
A drug-linker conjugate that couples the α-amanitin payload to a cleavable MC-VC-PABC-C6 linker for antibody-drug conjugate (ADC) research applications. The conjugate is designed to deliver α-amanitin selectively to target cells and acts as a potent inhibitor of RNA polymerase IIα.
  • Potent inhibitor of RNA polymerase IIα.
  • Designed for ADC development and preclinical research.
  • Available in microgram and milligram pack sizes; larger quantities available by quote.
  • Solid form; store sealed, away from moisture and light.
  • Stability in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-02426 Supplier Medchemexpress LLC Supplier No. HY1310891MG
Add to Cart
Add to Cart
Request a Quote
A drug-linker conjugate that couples the α-amanitin payload to a cleavable MC-VC-PABC-C6 linker for antibody-drug conjugate (ADC) research applications. The conjugate is designed to deliver α-amanitin selectively to target cells and acts as a potent inhibitor of RNA polymerase IIα.
  • Potent inhibitor of RNA polymerase IIα.
  • Designed for ADC development and preclinical research.
  • Available in microgram and milligram pack sizes; larger quantities available by quote.
  • Solid form; store sealed, away from moisture and light.
  • Stability in solvent: -80°C (6 months), -20°C (1 month).