Learn More
Medchemexpress LLC Mc-vc-pabc-c6-α-amanitin | 2922514-69-8 | 98.9% | 1616.79 g/mol | C74H105N17O22S | 500 UG
SDP
Catalog No. 5000203709
Encompass_Preferred
MC-VC-PABC-C6-α-Amanitin is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research that links the RNA polymerase II inhibitor α-amanitin to an MC-VC-PABC-C6 cleavable linker to enable targeted delivery of the toxin.
  • Used for ADC development and conjugation studies.
  • Contains α-amanitin as the cytotoxic payload linked via a valine-citrulline PABC C6 spacer.
  • High purity and characterized by LC-MS and RP-HPLC analytical data.
  • Available in small research quantities with recommended -20°C storage; in solvent stability guidance provided.
Catalog No. 50-002-03709 Supplier Medchemexpress LLC Supplier No. HY131089500UG
Add to Cart
Add to Cart
Request a Quote
MC-VC-PABC-C6-α-Amanitin is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research that links the RNA polymerase II inhibitor α-amanitin to an MC-VC-PABC-C6 cleavable linker to enable targeted delivery of the toxin.
  • Used for ADC development and conjugation studies.
  • Contains α-amanitin as the cytotoxic payload linked via a valine-citrulline PABC C6 spacer.
  • High purity and characterized by LC-MS and RP-HPLC analytical data.
  • Available in small research quantities with recommended -20°C storage; in solvent stability guidance provided.