Learn More
LGC GENOMICS LLC (Lucigen) MC1061 F- Electrocompetent Cells, 24 Rxns (DUOs)High efficiency F-control strain for phage display applications.
Catalog No. NC1999480
Encompass
Perfect for antibody phage display or peptide phage display library creationThe highest efficiency TG1 competent cells availableYour only source for electrocompetent SS320 and ER2738 cellsAmber suppressor and non-amber suppressor lines for maximum flexibilityHigh efficiency MC1061 F- cells for cloning or control reactionsCreate larger libraries; speed discoveryBulk custom dispensing available with all of our phage display cellsCompetent Cells include Control DNA and Recovery Medium and are packaged as DUOs (2 transformations per tube). Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.
Catalog No. NC1999480 Supplier LGC GENOMICS LLC Supplier No. 605142
Perfect for antibody phage display or peptide phage display library creationThe highest efficiency TG1 competent cells availableYour only source for electrocompetent SS320 and ER2738 cellsAmber suppressor and non-amber suppressor lines for maximum flexibilityHigh efficiency MC1061 F- cells for cloning or control reactionsCreate larger libraries; speed discoveryBulk custom dispensing available with all of our phage display cellsCompetent Cells include Control DNA and Recovery Medium and are packaged as DUOs (2 transformations per tube). Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.