Learn More
Enzo Life Sciences MC1293 (5mg)
SDP
Supplier:  Enzo Life Sciences ALX270344M005
Encompass_Preferred
HDAC1 inhibitor. Inhibitor of histone deacetylase 1 (HDAC1) (IC50 = 4.5µM) and maize histone deacetylase HD2 (IC50=1.9µM). Alternative name: 3-(4-Toluoyl-1-methyl-1H-2-pyrrolyl)-N-hydroxy-2-propenamide. Formula: C16H16N2O3. MW: 284.3. Purity: ≥97% (HPLC). Appearance: Off-white powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8491