Learn More
Medchemexpress LLC HY-16914 10mg Medchemexpress, MC1568 CAS:852475-26-4 Purity:>98%
SDP
Catalog No. 502081915
Encompass_Preferred
Medchemexpress, HY-16914 10mg MC1568 CAS:852475-26-4 MC1568 is a selective class II (IIa) histone deacetylas (HDAC II) inhibitor, used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1915 Supplier Medchemexpress LLC Supplier No. HY1691410MG
Medchemexpress, HY-16914 10mg MC1568 CAS:852475-26-4 MC1568 is a selective class II (IIa) histone deacetylas (HDAC II) inhibitor, used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.