Learn More
Apexbio Technology LLC MC1568 852475-26-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063995
Encompass_Preferred
MC1568 (CAS 852475-26-4) is a histone deacetylase (HDAC) inhibitor derived from the (aryloxopropenyl)pyrrolyl hydroxyamide chemical class It selectively targets class II HDAC enzymes particularly class IIa subtypes (HDAC4 HDAC5 HDAC7 HDAC9) inhibiting maize class II HDAC activity with an IC50 of 22 M MC1568 modulates muscle differentiation pathways by reducing MEF2D expression stabilizing HDAC-HDAC3-MEF2D complexes and preventing MEF2D acetylation in cultured myocytes It also interferes with differentiation signals mediated via RAR and PPAR nuclear receptors serving as a valuable tool in epigenetic and cellular differentiation studies
Catalog No. 50-000-63995 Supplier Apexbio Technology LLC Supplier No. A40945.1
MC1568 (CAS 852475-26-4) is a histone deacetylase (HDAC) inhibitor derived from the (aryloxopropenyl)pyrrolyl hydroxyamide chemical class It selectively targets class II HDAC enzymes particularly class IIa subtypes (HDAC4 HDAC5 HDAC7 HDAC9) inhibiting maize class II HDAC activity with an IC50 of 22 M MC1568 modulates muscle differentiation pathways by reducing MEF2D expression stabilizing HDAC-HDAC3-MEF2D complexes and preventing MEF2D acetylation in cultured myocytes It also interferes with differentiation signals mediated via RAR and PPAR nuclear receptors serving as a valuable tool in epigenetic and cellular differentiation studies