Learn More
Apexbio Technology LLC MC1568, 25mg. Cas: 852475-26-4 MFCD: MFCD16875423. Class II HDAC inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A409425
Encompass_Preferred
MC1568, a derivative of (Aryloxopropenyl)pyrrolyl hydroxyamide, is a novel, potent and specific inhibitor of class II histone deacetylase (HDAC) that exhibits strong inhibition against maize class II HDAC with 50% inhibition concentration IC50 value of 22 M. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4876
 Restricted Item