Learn More
Biomatik Corporation Mc1R Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11237
Encompass_Preferred
Mc1R Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MC1R. The Mc1R Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Melanocyte-Stimulating Hormone Receptor Protein (1-37Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: MC1R. Accession Number: Q01726. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5909