Learn More
TARGETMOL CHEMICALS INC MC2590 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T735155MG
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. MC2590 is a potent and selective histone deacetylase (HDAC) inhibitor that inhibits HDAC1-3 -6 -8 and -10 activities induces cell cycle arrest and promotes apoptosis. purity: 99%
Catalog No. 50-257-9207