Learn More
Aviva Systems Biology Corp MC2R ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP63649_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MC2R. It was validated on Western Blot. . Alternate names: ACTHR, MGC125798ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-288-042