Learn More
Organix Inc (MC3) DLIN-MC3-DMA, 1 g
SDP
Catalog No. NC1779924
Encompass
Dlin-MC3-DMA also known as MC3, is one of the most utilized cationic lipids that is used for making Lipid Nano-Particles (LNPs). These lipids, can form nanoparticles that can encapsulate and deliver siRNA, mRNA, DNA or small molecules into the cytoplasm and represent the most advanced delivery platforms for systemic administration of active molecules.
Catalog No. NC1779924 Supplier Organix Inc Supplier No. O8640 1G
Add to Cart
Add to Cart
Request a Quote
Dlin-MC3-DMA also known as MC3, is one of the most utilized cationic lipids that is used for making Lipid Nano-Particles (LNPs). These lipids, can form nanoparticles that can encapsulate and deliver siRNA, mRNA, DNA or small molecules into the cytoplasm and represent the most advanced delivery platforms for systemic administration of active molecules.