Learn More
Medchemexpress LLC HY-112587 10mg Medchemexpress, MC3482 CAS: Purity:>98%
SDP
Catalog No. 501969108
Encompass_Preferred
Medchemexpress, HY-112587 10mg MC3482 CAS: MC3482 is a specific sirtuin5 (SIRT5) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9108 Supplier Medchemexpress LLC Supplier No. HY11258710MG
Medchemexpress, HY-112587 10mg MC3482 CAS: MC3482 is a specific sirtuin5 (SIRT5) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.