Learn More
TARGETMOL CHEMICALS INC MC3482 5MG
SDP
Catalog No. 502581496
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MC3482 is a specific inhibitor of sirtuin5 (SIRT5).
Catalog No. 50-258-1496 Supplier TARGETMOL CHEMICALS INC Supplier No. T11962
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MC3482 is a specific inhibitor of sirtuin5 (SIRT5).