Learn More
Eurofins Discoverx PathHunter MC3R Total Internalization U2OS Cell Line, 2 vials. (93-0837C3)
Catalog No. 502001390
Encompass
The PathHunter MC3R Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of MC3R via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged MC3R and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the MC3R receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1390 Supplier Eurofins Discoverx Supplier No. 930837C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter MC3R Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of MC3R via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged MC3R and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the MC3R receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.