Learn More
ATCC Mus musculus
Supplier:  ATCC CRL2593/DEL
Encompass
MC3T3-E1 Subclone 4 preosteoblasts were isolated from the cloned but phenotypically heterogeneous MC3T3-E1 cell line. This subclone was selected for high osteoblast differentiation and mineralization after growth in medium containing ascorbic acid. This cell line is a good model for studying in vitro osteoblast differentiation, particularly ECM signaling. It's behavior is similar to primary calvarial osteoblasts.
Catalog No. 50-188-646FP