Learn More
ATCC MC3T3-E1 Subclone 4; Preosteoblast; Mouse (Mus musculus)
Catalog No. 502383383
Encompass
MC3T3-E1 Subclone 4 preosteoblasts were isolated from the cloned but phenotypically heterogeneous MC3T3-E1 cell line. This subclone was selected for high osteoblast differentiation and mineralization after growth in medium containing ascorbic acid. This cell line is a good model for studying in vitro osteoblast differentiation, particularly ECM signaling. It's behavior is similar to primary calvarial osteoblasts.
Catalog No. 50-238-3383 Supplier ATCC Supplier No. CRL2593
 Restricted Item
Add to Cart
Add to Cart
MC3T3-E1 Subclone 4 preosteoblasts were isolated from the cloned but phenotypically heterogeneous MC3T3-E1 cell line. This subclone was selected for high osteoblast differentiation and mineralization after growth in medium containing ascorbic acid. This cell line is a good model for studying in vitro osteoblast differentiation, particularly ECM signaling. It's behavior is similar to primary calvarial osteoblasts.