Learn More
Eurofins Discoverx PathHunter MC4R Bioassay Kit, 10-Plate. (93-0211Y3-00086)
Catalog No. 501999966
Encompass
The PathHunter MC4R Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. αMSH) based activation of MC4R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MC4R and EA-tagged β-Arrestin-2. Activation of MC4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-199-9966 Supplier Eurofins Discoverx Supplier No. 930211Y300086
Add to Cart
Add to Cart
Request a Quote
The PathHunter MC4R Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. αMSH) based activation of MC4R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MC4R and EA-tagged β-Arrestin-2. Activation of MC4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.