Learn More
Eurofins Discoverx ChemiScreen MC5 Melanocortin Receptor Membrane Preparation, 1 vial. (HTS155M)
Catalog No. 502005976
Encompass
The ChemiScreen MC5 Melanocortin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of MC5R surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5976 Supplier Eurofins Discoverx Supplier No. HTS155M
The ChemiScreen MC5 Melanocortin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of MC5R surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.