Learn More
ATCC MC57G; Fibrosarcoma; Mouse (Mus musculus)
Catalog No. 502381960
Encompass
MC57G is a cell line that was isolated from a mouse with fibrosarcoma. The tumor arose in the C57BL/6 mouse after treatment with methylcholanthrene. This cell line is the laboratory standard in laboratories for mouse viral immunity studies.
Catalog No. 50-238-1960 Supplier ATCC Supplier No. CRL2295
MC57G is a cell line that was isolated from a mouse with fibrosarcoma. The tumor arose in the C57BL/6 mouse after treatment with methylcholanthrene. This cell line is the laboratory standard in laboratories for mouse viral immunity studies.